Ammonium cerium(IV) nitrate ≥98.5% ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-5803-100G 93-5803-500G
100205-680EA 124.55 USD
100205-680 100210-496
Ammonium cerium(IV) nitrate ≥98.5% ACS
Ammonium cerium(IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MDL Number: MFCD00151121
CAS Number: 16774-21-3
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR