Cerium(III) acetylacetonate hydrate ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cerium(III) 2,4-pentanedionate hydrate

93-5802-25G 93-5802-100G
100196-776EA 250.52 USD
100196-776 100208-728
Cerium(III) acetylacetonate hydrate ≥99.9% (Ce basis)
Cerium(III) acetylacetonate hydrate
Formula: C₁₅H₂₁CeO₆·xH₂O
MW: 437.44 g/mol (anhy)
MDL Number: MFCD00150164
CAS Number: 206996-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR