Lanthanum(III) bromide heptahydrate ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS

93-5730-25G 93-5730-5G
100204-384EA 107.57 USD
100204-384 100205-674
Lanthanum(III) bromide heptahydrate ≥99.9% (La basis)
Lanthanum(III) bromide heptahydrate
MDL Number: MFCD00150247
CAS Number: 13465-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR