Lanthanum(III) phosphate hydrate
Supplier: STREM CHEMICALS INC MS
93-5717-100G
93-5717-25G
100202-214EA
396.31
USD
100202-214
100204-734
Lanthanum(III) phosphate hydrate
Lanthanum(III) phosphate hydrate
Formula:
LaPO₄.xH₂O MW: 233.877 g/mol (anhy) |
MDL Number:
MFCD00150248 CAS Number: 14913-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...