Lanthanum(III) carbonate hydrate ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS

93-5707-500G 93-5707-100G
100209-160EA 491.14 USD
100209-160 100200-588
Lanthanum(III) carbonate hydrate ≥99.9% (La basis)
Lanthanum(III) carbonate hydrate
Formula: La₂(CO₃)₃.xH₂O
MW: 457.85 g/mol (anhy)
Density: 2.6…2.7
MDL Number: MFCD00011069
CAS Number: 54451-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR