Lanthanum(III) 2,4-pentanedionate hydrate ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lanthanum(III) acetylacetonate hydrate, Tris(2,4-pentanedionato)lanthanum(III) hydrate, Acetylacetone Lanthanum(III) hydrate
93-5702-25G
93-5702-100G
100196-766EA
97.66
USD
100196-766
100199-632
Lanthanum(III) 2,4-pentanedionate hydrate ≥99.9% (La basis)
Lanthanum(III) 2,4-pentanedionate hydrate
Formula:
C₁₅H₂₁LaO₆·xH₂O MW: 436.24 g/mol (anhy) |
MDL Number:
MFCD00149057 CAS Number: 64424-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...