Lanthanum triacetate hydrate ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lanthanum(III) acetate hydrate
93-5701-100G
93-5701-500G
100204-380EA
94.83
USD
100204-380
100200-586
Lanthanum triacetate hydrate ≥99.9% (La basis)
Lanthanum triacetate hydrate
Formula:
C₆H₉LaO₆·H₂O MW: 334.053 g/mol |
MDL Number:
MFCD00150118 CAS Number: 100587-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...