Barium acetylacetonate hydrate

Supplier: STREM CHEMICALS INC MS
Danger

93-5657-10G 93-5657-50G 93-5657-250G
100204-378EA 45.29 USD
100204-378 100205-666 100202-462
Barium acetylacetonate hydrate
Barium acetylacetonate hydrate
Formula: C₁₀H₁₄BaO₄·xH₂O
Melting Pt: > 320 °C
MDL Number: MFCD00013484
CAS Number: 12084-29-6
UN: 1564
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR