Barium-2-ethylhexanoate ∼30% in xylene (7-10% Ba)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5626-100G
93-5626-25G
100207-110EA
229.29
USD
100207-110
100211-274
Barium-2-ethylhexanoate ∼30% in xylene (7-10% Ba)
Barium 2-ethylhexanoate
Formula:
C₁₆H₃₀BaO₄ Flash Pt: 85 °F |
MDL Number:
MFCD00058696 CAS Number: 2457-01-4 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...