Molecular sieve 13X (1.0 nm, 10 Å), pellets 1/16''

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Linde 13X

93-5412-250G 93-5412-1000G
100196-746EA 65.11 USD
100196-746 100199-614
Molecular sieve 13X (1.0 nm, 10 Å), pellets 1/16''
Molecular sieve 13X (1.0 nm, 10 Å)

Molecular Sieve

MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR