Molecular sieve 13X (1.0 nm, 10 Å), Powder -600 mesh

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Linde 13X

93-5411-250G 93-5411-1000G
100197-896EA 66.52 USD
100197-896 100199-612
Molecular sieve 13X (1.0 nm, 10 Å), Powder -600 mesh
Molecular sieve 13X (1.0 nm, 10 Å)

Molecular Sieve

MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR