Molecular sieve A5 (0.5 nm, 5 Å), pellets 1/8''

Supplier: STREM CHEMICALS INC MS

93-5410-250G 93-5410-1000G
100196-744EA 66.52 USD
100196-744 100199-610
Molecular sieve A5 (0.5 nm, 5 Å), pellets 1/8''
Molecular sieve A5 (0.5 nm, 5 Å)

Molecular Sieve

MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR