Molecular sieve A5 (0.5 nm, 5 Å), pellets 1/16''

Supplier: STREM CHEMICALS INC MS

93-5409-1000G 93-5409-250G
100199-608EA 159.94 USD
100199-608 100196-742
Molecular sieve A5 (0.5 nm, 5 Å), pellets 1/16''
Molecular sieve A5 (0.5 nm, 5 Å)

Molecular Sieve

MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR