Molecular sieve A5 (0.5 nm, 5 Å), Powder -600 mesh
Supplier: STREM CHEMICALS INC MS
Synonyms:
Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
93-5408-500G
93-5408-2000G
100205-648EA
72.18
USD
100205-648
100209-150
Molecular sieve A5 (0.5 nm, 5 Å), Powder -600 mesh
Molecular sieve A5 (0.5 nm, 5 Å)
Molecular Sieve
MDL Number:
MFCD00131613 CAS Number: 69912-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...