Triphenylantimony dibromide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-5125-1G 93-5125-5G
100205-638EA 99.08 USD
100205-638 100203-508
Triphenylantimony dibromide ≥98%
Triphenylantimony dibromide
Formula: C₁₈H₁₅Br₂Sb
MDL Number: MFCD00045008
CAS Number: 1538-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR