Triphenylantimony dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-5124-2G 93-5124-10G
100199-130EA 46.71 USD
100199-130 100203-506
Triphenylantimony dichloride ≥99%
Triphenylantimony dichloride
Formula: C₁₈H₁₅Cl₂Sb
Melting Pt: 143…145 °C
MDL Number: MFCD00013598
CAS Number: 594-31-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR