Antimony triacetate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Antimony acetate, Anitmony (III) acetate, Triacetoxystibine, Antimony(III) acetate, Antimony triacetate, Acetic acid antimony (III) salt
93-5111-250G
93-5111-50G
100197-252EA
325.54
USD
100197-252
100207-866
Antimony triacetate ≥97%
Antimony triacetate
Formula:
(H₃CCOO)₃Sb MW: 298.89 g/mol |
MDL Number:
MFCD00014974 CAS Number: 6923-52-0 UN: 1549 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...