Tin(II) phthalocyanine

Supplier: STREM CHEMICALS INC MS

93-5072-250MG 93-5072-1G
100205-628EA 77.85 USD
100205-628 100205-888
Tin(II) phthalocyanine
Tin (II) phthalocyanine
Formula: C₃₂H₁₆N₈Sn
MDL Number: MFCD00058867
CAS Number: 15304-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR