Indium(III) acetate ≥99.99% (In basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4936-25G
93-4936-5G
100207-092EA
270.34
USD
100207-092
100202-166
Indium(III) acetate ≥99.99% (In basis), PURATREM
Indium(III) acetate
Formula:
C₂H₄InO₂ MW: 174.87 g/mol |
MDL Number:
MFCD00054347 CAS Number: 25114-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...