Indium(III) chloride tetrahydrate ≥99.99% (In basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-4931-5G 93-4931-25G
100196-700EA 69.35 USD
100196-700 100207-480
Indium(III) chloride tetrahydrate ≥99.99% (In basis), PURATREM
Indium (III) chloride tetrahydrate
Formula: Cl₃In·4H₂O
MDL Number: MFCD00149746
CAS Number: 22519-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR