Indium(III) perchlorate octahydrate ≥99.9% (In basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-4925-25G 93-4925-5G
100202-162EA 389.23 USD
100202-162 100202-164
Indium(III) perchlorate octahydrate ≥99.9% (In basis)
Indium(III) perchlorate octahydrate
Formula: In(ClO₄)₃.8H₂O
Boiling Pt: 200 °C (dec.)
Melting Pt: ~80 °C
MDL Number: MFCD00150241
CAS Number: 13465-15-1
UN: 1481
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR