Cadmium(II) acetylacetonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4802-25G
93-4802-100G
100201-116EA
230.71
USD
100201-116
100196-984
Cadmium(II) acetylacetonate ≥98%
Cadmium(II) acetylacetonate
Formula:
C₁₀H₁₄CdO₄ Melting Pt: Dec. |
MDL Number:
MFCD00040412 CAS Number: 14689-45-3 UN: 2570 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...