Silver perchlorate monohydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4749-10G
93-4749-50G
100200-582EA
162.77
USD
100200-582
100209-124
Silver perchlorate monohydrate ≥99%
Silver perchlorate monohydrate
Formula:
AgClO₄·1H₂O MW: 225.33 |
MDL Number:
MFCD00149128 CAS Number: 14242-05-8 UN: 1481 ADR: 5.1 II Merck Index: 12,08667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...