Silver cyclohexanebutyrate standard for AAS

Supplier: STREM CHEMICALS INC MS
Warning

93-4709-5G 93-4709-1G
100206-034EA 148.62 USD
100206-034 100209-118
Silver cyclohexanebutyrate standard for AAS
Silver cyclohexanebutyrate
Formula: C₁₀H₁₇AgO₂
MDL Number: MFCD00036396
CAS Number: 62638-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR