trans-Diamminedichloropalladium(II) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: trans-Dichlorodiammine palladium(II), Dichlorodiamminepalladium, trans-Diammine dichloropalladium

93-4614-5G 93-4614-1G
100196-670EA 750.15 USD
100196-670 100196-668
trans-Diamminedichloropalladium(II) ≥99%
trans-Diamminedichloropalladium(II)
Formula: Cl₂H₆N₂Pd
MW: 211.39 g/mol
MDL Number: MFCD00011621
CAS Number: 13782-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR