Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tris(triphenylphosphoranyl)ruthenium(V) chloride hydride
93-4401-1G
93-4401-250MG
100208-722EA
253.35
USD
100208-722
100196-664
Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct ≥98%
Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct
Formula:
C₅₄H₄₉ClP₃Ru MW: 927.4112 g/mol |
MDL Number:
MFCD04971968 CAS Number: 55102-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...