12-Molybdosilicic acid hydrate

Supplier: STREM CHEMICALS INC MS
Warning

93-4216-10G 93-4216-50G
100207-844EA 86.34 USD
100207-844 100208-720
12-Molybdosilicic acid hydrate
12-Molybdosilicic acid hydrate
MDL Number: MFCD00150575
CAS Number: 11089-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR