Molybdenum disilicide ≥99% (Mo basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Molybdenum silicide

93-4212-100G 93-4212-25G
100200-764EA 318.46 USD
100200-764 100205-576
Molybdenum disilicide ≥99% (Mo basis)
Molybdenum disilicide
Formula: MoSi₂
MW: 152.11 g/mol
Density: 6.31
MDL Number: MFCD00151535
CAS Number: 12136-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR