Molybdenum(VI) tetrachloride oxide ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

93-4209-25G 93-4209-5G
100202-704EA 1738.09 USD
100202-704 100210-404
Molybdenum(VI) tetrachloride oxide ≥97%
Molybdenum(VI) tetrachloride oxide
Melting Pt: 100…101 °C
MDL Number: MFCD00014222
CAS Number: 13814-75-0
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR