Potassium niobate ≥99.999% (Nb basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-4117-50G 93-4117-10G
100199-566EA 754.4 USD
100199-566 100202-126
Potassium niobate ≥99.999% (Nb basis), PURATREM
Potassium niobate
Formula: KNbO₃
MW: 180.002 g/mol
MDL Number: MFCD00049651
CAS Number: 12030-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR