Zirconium(IV) ethoxide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-4043-25G 93-4043-5G
100205-880EA 401.97 USD
100205-880 100200-762
Zirconium(IV) ethoxide ≥99%
Zirconium(IV) ethoxide

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₈H₂₀O₄Zr
MW: 271.47 g/mol
Melting Pt: 171…173 °C
MDL Number: MFCD00015654
CAS Number: 18267-08-8
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR