Tetrakis(diethylamino)zirconium(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetrakis(diethylamino)zirconium

93-4040-5G 93-4040-25G
100197-850EA 223.63 USD
100197-850 100197-848
Tetrakis(diethylamino)zirconium(IV) ≥99%
Tetrakis(diethylamino)zirconium(IV)

Metal Alkylamides, Volatile Precursors for CVD

Formula: C₁₆H₄₀N₄Zr
MW: 379.74 g/mol
Boiling Pt: 112 °C (0.1 mmHg)
Density: 1.026
MDL Number: MFCD00015649
CAS Number: 13801-49-5
UN: 3399
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR