Zirconium(IV) sulfide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-4033-10G 93-4033-2G
100200-568EA 1463.51 USD
100200-568 100200-570
Zirconium(IV) sulfide ≥99%
Zirconium(IV) sulfide
Melting Pt: 1550 °C
Density: 3.87
MDL Number: MFCD00054116
CAS Number: 12039-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR