Zirconium silicate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-4019-1000G 93-4019-250G
100201-110EA 167.02 USD
100201-110 100208-560
Zirconium silicate ≥98%
Zirconium silicate
Formula: O₄SiZr
MW: 183.305 g/mol
Melting Pt: 2550 °C
Density: 4.56
MDL Number: MFCD00085353
CAS Number: 10101-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR