Zirconium(IV) Propoxide (23-28% free alcohol)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Zirconium(IV) Tetrapropoxide, Zirconium(IV) n-propoxide
93-4017-1000G
93-4017-250G
100200-760EA
283.08
USD
100200-760
100205-560
Zirconium(IV) Propoxide (23-28% free alcohol)
Zirconium(IV) Propoxide
Alkoxide
Formula:
C₁₂H₂₈O₄Zr MW: 327.58 g/mol Density: 1.05 Flash Pt: 83 °F |
MDL Number:
MFCD00015307 CAS Number: 23519-77-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...