Zirconium(IV) carbonate, basic hydrate

Supplier: STREM CHEMICALS INC MS
Warning

93-4007-500G 93-4007-2000G
100197-846EA 93.42 USD
100197-846 100211-256
Zirconium(IV) carbonate, basic hydrate
Zirconium(IV) carbonate, basic hydrate
MDL Number: MFCD00172265
CAS Number: 12671-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR