Zirconium carbide ≥99% (Zr basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-4006-100G 93-4006-25G
100210-880EA 298.65 USD
100210-880 100202-114
Zirconium carbide ≥99% (Zr basis)
Zirconium carbide
Formula: CZr
MW: 103.235 g/mol
Boiling Pt: 5100 °C
Melting Pt: 3540 °C
Density: 6.73
MDL Number: MFCD00049640
CAS Number: 12070-14-3
UN: 3178
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR