Zirconium(IV) butoxide 76 - 80% in n-butanol

Supplier: STREM CHEMICALS INC MS
Danger

93-4003-250G 93-4003-1000G 93-4003-50G
100200-142EA 77.85 USD
100200-142 100207-056 100199-096
Zirconium(IV) butoxide 76 - 80% in n-butanol
Zirconium(IV) butoxide

Alkoxide

Formula: C₁₆H₃₆O₄Zr
Density: 1.063
Flash Pt: 101 °F
MDL Number: MFCD00048765
CAS Number: 1071-76-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR