Yttrium(III) isopropoxide

Supplier: STREM CHEMICALS INC MS
Danger

93-3932-1G 93-3932-25G 93-3932-5G
100199-088EA 83.51 USD
100199-088 100199-090 100199-092
Yttrium(III) isopropoxide
Yttrium(III) isopropoxide

Alkoxide

Formula: C₉H₂₁O₃Y
MW: 266.17 g/mol
MDL Number: MFCD00015641
CAS Number: 2172-12-5
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR