Yttrium(III) oxalate hydrate ≥99.9% (Y basis)

Supplier: STREM CHEMICALS INC MS

93-3931-25G 93-3931-100G
100196-646EA 72.18 USD
100196-646 100202-112
Yttrium(III) oxalate hydrate ≥99.9% (Y basis)
Yttrium(III) oxalate hydrate
Formula: C₆O₁₂Y₂·xH₂O
MW: 441.87 g/mol (anhy)
MDL Number: MFCD00150500
CAS Number: 13266-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR