Yttrium(III) sulfate octahydrate ≥99.9% (Y basis)

Supplier: STREM CHEMICALS INC MS

93-3916-25G 93-3916-100G
100208-558EA 65.11 USD
100208-558 100198-374
Yttrium(III) sulfate octahydrate ≥99.9% (Y basis)
Yttrium(III) sulfate octahydrate
Formula: H₁₆O₂₀S₃Y₂
MW: 610.1 g/mol
Density: 2.558
MDL Number: MFCD00149946
CAS Number: 7446-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR