Strontium zirconium oxide ≥95%

Supplier: STREM CHEMICALS INC MS

93-3826-1000G 93-3826-250G
100197-836EA 435.94 USD
100197-836 100209-108
Strontium zirconium oxide ≥95%
Strontium zirconium oxide
Formula: SrZrO₃
MDL Number: MFCD00049555
CAS Number: 12036-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR