Cacodylic acid (dimethylarsinic acid) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-3318-10G
93-3318-50G
100196-628EA
145.78
USD
100196-628
100198-362
Cacodylic acid (dimethylarsinic acid) ≥98%
Cacodylic acid (dimethylarsinic acid)
Formula:
(CH₃)₂As(O)OH Melting Pt: 200 °C |
MDL Number:
MFCD00002095 CAS Number: 75-60-5 UN: 1572 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...