Germanium(II) sulfide ≥99.99% (Ge basis) (contains 1-5% germanium metal), PURATREM

Supplier: STREM CHEMICALS INC MS

93-3209-2G 93-3209-10G
100205-876EA 199.57 USD
100205-876 100200-336
Germanium(II) sulfide ≥99.99% (Ge basis) (contains 1-5% germanium metal), PURATREM
Germanium(II) sulfide
Melting Pt: 530 °C
MDL Number: MFCD00135539
CAS Number: 12025-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR