Ethylgermanium trichloride ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

93-3203-1G 93-3203-5G
100199-072EA 94.83 USD
100199-072 100207-292
Ethylgermanium trichloride ≥97%
Ethylgermanium trichloride
Boiling Pt: 144 °C
Melting Pt: –33 °C
Density: 1,604
MDL Number: MFCD00013586
CAS Number: 993-42-0
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR