Zinc 2-ethylhexanoate ≥80% in mineral spirits (14-15% Zn)

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc(II) 2-ethylhexanoate

93-3038-500G 93-3038-100G
100202-076EA 382.15 USD
100202-076 100204-306
Zinc 2-ethylhexanoate ≥80% in mineral spirits (14-15% Zn)
Zinc 2-ethylhexanoate
Formula: C₁₆H₃₀O₄Zn
MW: 351.792 g/mol
Density: 1.07
Flash Pt: 104 °F
MDL Number: MFCD00053316
CAS Number: 136-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR