Zinc molybdate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc molybdenum oxide

93-3036-10G 93-3036-2G
100203-442EA 308.55 USD
100203-442 100205-512
Zinc molybdate ≥98%
Zinc molybdate
Formula: MoO₄Zn
MDL Number: MFCD00049622
CAS Number: 13767-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR