Zinc phosphate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

93-3020-500G 93-3020-2000G
100205-510EA 86.34 USD
100205-510 100210-344
Zinc phosphate hydrate
Zinc phosphate hydrate
Formula: O₈P₂Zn₃·xH₂O
MW: 386.08 g/mol (anhy)
Density: 3.04
MDL Number: MFCD00036282
CAS Number: 7779-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR