Zinc tetrafluoroborate hydrate ≥98%

Supplier: STREM CHEMICALS INC MS

93-3010-100G 93-3010-25G
100196-616EA 202.4 USD
100196-616 100207-028
Zinc tetrafluoroborate hydrate ≥98%
Zinc tetrafluoroborate hydrate
Formula: Zn(BF₄)₂·xH₂O
MW: 238.98 g/mol (anhy)
MDL Number: MFCD00150371
CAS Number: 27860-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR