Copper(II) trifluoroacetylacetonate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Copper(ii)trifluoroacetylacetonate
93-2966-25G
93-2966-5G
100197-808EA
338.28
USD
100197-808
100205-498
Copper(II) trifluoroacetylacetonate ≥97%
Copper(II) trifluoroacetylacetonate
Metal Beta-diketonates, Volatile Precursors for CVD
Boiling Pt:
260 °C/ 140 °C (0.1 mmHg) (dec./ subl.) Melting Pt: 194…196 °C |
MDL Number:
MFCD00042511 CAS Number: 14324-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...